PHARMED Tubing
Advantages:
- Great for tissue and cell worknontoxic and nonhemolytic.
- Long service life minimizes risk of fluid exposure; reduces tubing costs and pump downtime.
- Opaque to UV and visible light to protect light-sensitive fluids.
- Heat sealable, bondable, and formable.
- Extremely low gas permeability.
Limitations:
- Potential leaching of USP mineral oil or blend material.
Application Suitability:
Acids: | Good |
Alkalies: | Good |
Organic solvents: | Not recommended |
Pressure: | Good |
Vacuum: | Excellent |
Viscous fluids: | Good |
Sterile fluids: | Excellent |
Physical Characteristics and Compostion:
- Thermoplastic elastomer.
- Polypropylene-based material with USP mineral oil.
- Excellent tensile strength.
- Firm (stiff) material.
- Opaque, beige.
Temperature Range:
-75 to 275°F (-59 to 135°C)
Meets Classifications:
- USP Class VI
- FDA 21 CFR 177.2600
- NSF listed (Standard 51)
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2: | 1200 | |
H2: | - | |
O2: | 200 | |
N2: | 80 |
Cleaning/Sterilization:
- Sterilize by ethylene oxide (ETO), autoclave or gamma irradiation up to 2.5 Mrad.
- Repeated autoclaving will not affect overall life.
Last Updated: 10/15/18