C-FLEX (50 A) Tubing
Advantages:
- Physical properties similar to silicone with chemical compatibility of TYGON®.
- Inexpensive.
- Excellent biocompatibility.
- Smooth bore.
- Nontoxic, no leachable plasticizers.
- Lower gas permeability than silicone.
- Use with many acids and alkalies.
Limitations:
- Not recommended for use with oils.
- Moderate pumping life.
Application Suitability:
Acids: | Good |
---|---|
Alkalies: | Good |
Organic solvents: | Not recommended |
Pressure: | Fair |
Vacuum: | Good |
Viscous fluids: | Fair |
Sterile fluids: | Excellent |
Physical Characteristics and Compostion:
- Thermoplastic elastomer.
- Styrene-ethylene-butylene modified block copolymer with silicone oil.
- Excellent tensile and tear strength.
- Soft material.
- Opaque, white.
Temperature Range:
-100 to 275°F (-73 to 135°C)
Meets Classifications:
- USP Class VI
- FDA 21 CFR 177.2600
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2: | - |
---|---|
H2: | - |
O2: | 150 |
N2: | - |
Cleaning/Sterilization:
- Sterilize by ethylene oxide (ETO), autoclave or gamma irradiation.
Last Updated: 10/15/18