Stampa…
IPTG (Isopropil-β-D-tiogalattopiranoside) ≥99% per biologia molecolare
Fornitore: G Biosciences
Synonyms: IPTG
R036 RC-062 RC-063
R036EA 380 EUR
R036 RC-062 RC-063
IPTG (Isopropil-β-D-tiogalattopiranoside) ≥99% per biologia molecolare
IPTG (Isopropil-β-D-tiogalactopiranoside)
IPTG is a molecular mimic of allolactose that initiates transcription of the lac operon.
  • Blue/white screening
  • Induction of lac operon for protein expression
The key feature of IPTG is that, unlike allolactose, is not hydrolyzed by ß-galactosidase, therefore its concentration remains constant during an experiment. IPTG is therefore able to induce protein expression where the gene is under the control of the lac operator, when used as an effective inducer in the concentration range of 100 μM to 1.5 mM.
Formula: C₉H₁₈O₅S
Peso molecolare: 238,31 g/mol
Punto di ebollizione: 402 °C (1013 hPa)
Punto di fusione: 109,5…110.5 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0