Stampa…
Dog recombinant IL-4 (from E. coli)
Fornitore: Shenandoah Biotechnology
400-09AF-5UG 400-09AF-25UG 400-09AF-100UG 400-09AF-500UG 400-09AF-1MG
SHBT400-09AF-5UGEA 75.3 EUR
SHBT400-09AF-5UG SHBT400-09AF-25UG SHBT400-09AF-100UG SHBT400-09AF-500UG SHBT400-09AF-1MG
Dog recombinant IL-4 (from E. coli)
Proteine e Peptidi
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. In a positive feedback loop 2 T helper cells subsequently produce more Interleukin 4 (IL-4).
  • Animal-free
  • Produced from genetically modified E.coli
  • Monomeric protein, containing 109 amino acids
IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
Informazioni per l'ordine: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Attenzione: This product is for research use only and is not for use in humans.