Stampa…

Anti-CX3CR1 Rabbit Polyclonal Antibody

Fornitore: ProSci Inc.
2093 2093-0.02MG
PRSI2093EA 435 EUR
PRSI2093 PRSI2093-0.02MG
Anti-CX3CR1 Rabbit Polyclonal Antibody
Anticorpi
CX3CR1 Antibody: CX3CR1 is one of the chemokine receptors that are required as coreceptors for HIV infection. The genes encoding human, murine, and rat CX3CR1 were cloned and designated V28 and CMKBRL1, CX3CR1, and RBS11, respectively. The encoded seven transmembrane protein was recently identified as the receptor for a novel transmembrane molecule, fractalkine, and renamed CX3CR1. Recently, CX3CR1 was found to serve as a coreceptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by fractokine. CX3CR1 mediates leukocyte migration and adhesion. CX3CR1 isMore Product Information
Visualizza i codici

Specifiche

Simbolo dell'antigene CX3CR1
Tipo di anticorpi Chemokine (C-X3-C motif) receptor 1
Environmentally Preferable
Sinomini antigene CMKBRL1|V28|CCRL1|CMKDR1|GPR13|GPRV28
Clonalità Policlonale
Coniugazione Non coniugato
Reattività Human,Rat,Mouse
Ospite Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
ImmunoChimica Yes
ImmunoFluorescenza Yes
Citometria a flusso Yes
Immunogeno CX3CR1 antibody was raised against a peptide corresponding to amino acids 2 to 21 of human CX3CR1. The sequence differs from those of mouse and rat CX3CR1 by four amino acids.
Purificazione Affinity chromatography purified via peptide column
Tampone di conservazione PBS containing 0.02% sodium azide
Applicazioni testate Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR