Stampa…
DNA ladders, GeneRuler™ and O'GeneRuler™
Fornitore: Thermo Fisher Scientific
SM0371 SM0372 SM0373 SM0241 SM0242 SM0243 SM0321 SM0322 SM0323 SM0311 SM0312 SM0313 SM0331 SM0332 SM0333 SM1213 SM1211 SM1331 SM1332 SM1333 SM1334 SM1351 SM1352 SM1353 SM1212 SM0244 SM0314 SM0324 SM0334 SM1191 SM1192 SM1193 SM1551 SM1552 SM1553
SM0371EA 80.75 EUR
SM0371 SM0372 SM0373 SM0241 SM0242 SM0243 SM0321 SM0322 SM0323 SM0311 SM0312 SM0313 SM0331 SM0332 SM0333 SM1213 SM1211 SM1331 SM1332 SM1333 SM1334 SM1351 SM1352 SM1353 SM1212 SM0244 SM0314 SM0324 SM0334 SM1191 SM1192 SM1193 SM1551 SM1552 SM1553
DNA ladders, GeneRuler™ and O'GeneRuler™
Reagenti per elettroforesi Marker di pesi molecolari
Thermo Scientific™ GeneRuler™ and O'GeneRuler™ DNA ladders are mixtures of chromatography-purified individual DNA fragments. The GeneRuler™ line includes the most popular ladders, such as the 100 bp and 1 kb DNA Ladder. GeneRuler™ DNA ladders are available in two formats: Conventional (provided in TE buffer) and a ready-to-use format (premixed with 6X DNA Loading Dye which contains bromophenol blue and xylene cyanol FF). O'GeneRuler™ DNA ladders are another ready-to-use version of GeneRuler™ DNA ladders that are premixed with 6X Orange DNA Loading Dye.
  • Universal ladders for DNA sizing and quantification—cover broad range of DNA molecular weights and provide DNA amount per band
  • Sharp bands, bright reference bands
  • Supplied with loading dye for sample DNA
  • Ready-to-use ladders are stable for 6 months at room temperature
GeneRuler™ 1 kb, 100 bp, and 50 bp DNA ladders are recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels. GeneRuler™ DNA low range and high range ladders are best for sizing and approximate quantification of double-stranded DNA at low and high DNA size ranges.
O’GeneRuler™ DNA ladders are premixed with 6X Orange DNA Loading Dye, which contains xylene cyanol FF and orange G. In a 1% agarose gel orange G dye migrates at 50 bp. Therefore, O’GeneRuler™ DNA ladders are ideal to use when visualisation of small DNA fragments is important.
Attenzione: Note: GeneRuler™ DNA Ladders will be upgraded in April 2018. The upgraded product will arrive either premixed or provided with TriTrack loading buffer containing three colour tracking dyes. This allows monitoring of band separation from a wide range of DNA fragments. This upgrade will not affect the DNA ladder’s banding pattern or its performance. During the transition it is possible that both configurations will be shipped for a limited time.