Stampa…
Human Recombinant Acylphosphate Phosphohydrolase 2 (from E. coli)
Fornitore: ProSci Inc.
92-307
PRSI92-307EA 588 EUR
PRSI92-307
Human Recombinant Acylphosphate Phosphohydrolase 2 (from E. coli)
Enzimi
ACYP2, also known as Acylphosphatase-2, Acylphosphatase, muscle type isozyme, Acylphosphate phosphohydrolase 2, is a protein which belongs to the acylphosphatase family.ACYP2 contains one acylphosphatase-like domain.
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Muscle acylphosphatase and erythrocyte acylphosphatase have been isolated.ACYP2 plays an important role in hydrolyzing the phosphoenzyme intermediate of different membrane pumps. Recombinant human ACYP2 protein, fused to GST-tag at N-terminus, was expressed in E. coli.