Stampa…
Human Recombinant Sulfotransferase 2B1 (from E. coli)
Fornitore: ProSci Inc.
92-053
PRSI92-053EA 588 EUR
PRSI92-053
Human Recombinant Sulfotransferase 2B1 (from E. coli)
Enzimi
Sulfotransferase Family Cytosolic 2B Member 1(SULT2B1) is a member of the Sulfotransferase 1 family.
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
SULT2B1 is highly expressed in the placenta, prostate, and trachea with lower expression levels in the small intestine and lung. SULT2B1 catalyses the sulfate conjugation of numerous hormones, neurotransmitters, drugs, and xenobiotic compounds. Sulfonation enhances the water solubility of molecules, and therefore their renal excretion, however SULT2B1 can also result in bioactivation to form active metabolites.