Stampa…
Human recombinant PPP1CC (from E. coli)
Fornitore: ProSci Inc.
92-040
PRSI92-040EA 982 EUR
PRSI92-040
Human recombinant PPP1CC (from E. coli)
Proteine e Peptidi
Serine/Threonine-Protein Phosphatase PP1-Υ Catalytic Subunit (PPP1CC) is a member of the PPP phosphatase family. It is essential for cell division, participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1CC colocalizes with SPZ1 in the nucleus, with URI1 at mitochondrion, Rapidly exchanges between the nucleolar, nucleoplasmic and cytoplasmic compartments. As a cofactor, PPP1CC binds one iron ion and one manganese ion per subunit.. In addition, PPP1CC may play an important role in dephosphorylating substrates such as the postsynaptic density-associated Ca2+/calmodulin dependent protein kinase II.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N, C-6 His tag