Stampa…
Anti-ZNF521 Rabbit Polyclonal Antibody
Fornitore: ProSci Inc.
6065
PRSI6065EA 435 EUR
PRSI6065
Anti-ZNF521 Rabbit Polyclonal Antibody
Anticorpi
ZNF521 Antibody: The zinc finger protein 521 (ZNF521) is a transcription factor containing an N-terminal transcriptional repressor motif and 30 zinc finger domains. It plays a role in both erythroid cell and osteoblast differentiation during development, inhibiting the activities of early B-cell factor 1 (EBF1) in erythroid cells and Runx2 in osteoblast precursors. ZFP521 binds to both Runx2 and histone deacetylase 3 (HDAC3), promotes their association and antagonizes Runx2 transcriptional activity in a HDAC3-dependent manner, thereby regulating osteoblast differentiation, skeletalMore Product Information