Stampa…

Anti-LY6G Rat Monoclonal Antibody [clone: Nimp-R14]

Fornitore: ProSci Inc.
36-156
PRSI36-156EA 567 EUR
PRSI36-156
Anti-LY6G Rat Monoclonal Antibody [clone: Nimp-R14]
Anticorpi
Neutrophils are key components of the inflammatory response and as such contribute to the killing of microorganisms. In addition, recent evidence suggests their involvement in the development of the immune response. The Nimp-R14 mAb displays features of an optimal reagent allowing the selective depletion of mouse neutrophils in vivo.

Type: Primary
Antigen: LY6G
Clonality: Monoclonal
Clone: Nimp-R14
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR