Stampa…
Direct Coupled 303 Stainless Steel Rotary Vane Pumps
Fornitore: Avantor Fluid Handling
79411-10
MFLX79411-10EA 1320 EUR
MFLX79411-10
Direct Coupled 303 Stainless Steel Rotary Vane Pumps
Pompe Pompe con girante flessibile/rotativa/a rulli
Ideal for high pressure applications—up to 240 psi.
  • Pulseless flow at pressures up to 240 psi
  • Built-in bypass relief valve protects your pump from damage
  • Select from economical brass or corrosion-resistant 303 stainless steel bodied pumps
Power cord not included.