Stampa…
High-pressure, 303 stainless steel rotary vane pump, 2.3 GPM, 115/230 VAC
Fornitore: Avantor Fluid Handling
VWR Item#MFLX79411-10
Your Organization's Price:
Ideal for high pressure applications—up to 240 psi.
  • Pulseless flow at pressures up to 240 psi
  • Built-in bypass relief valve protects your pump from damage
  • Select from economical brass or corrosion-resistant 303 stainless steel bodied pumps
Power cord not included.