Stampa…
Filtered balance stations, XPert™
Fornitore: LABCONCO
3940220 3940421 3940221 3946522 3946622 3946542 3940321 3940420
LBCP3940220EA 0 EUR
LBCP3940220 LBCP3940421 LBCP3940221 LBCP3946522 LBCP3946622 LBCP3946542 LBCP3940321 LBCP3940420
Filtered balance stations, XPert™
Cappe e involucri Cappe di estrazione
XPert™ filtered balance stations are vented balance enclosures designed to protect the user during powder weighing operations. The built-in HEPA filter, mounted in the upper plenum, removes particulates, so that clean air is exhausted to the atmosphere. XPert® filtered balance station is 'stationary' because ducting to an outside blower or house exhaust system is required. Powder weighing applications found in pharmaceutical research and other industries are ideal for these enclosures. Interior dimensions accommodate large micro and analytical balances and containment-enhancing featuresMore Product Information