Stampa…
Ductless laboratory fume hood, benchtop, Basic™ 47
Fornitore: LABCONCO
2247401 2247301 2246601 2246602 2247501
LBCP2247401EA 0 EUR
LBCP2247401 LBCP2247301 LBCP2246601 LBCP2246602 LBCP2247501
Ductless laboratory fume hood, benchtop, Basic™ 47
Cappe e involucri Cappe per fumi, senza canalizzazione
Basic Laboratory Hoods are designed specifically for laboratories, classrooms and light duty applications where safe ventilation is required. Constructed of epoxy-coated steel, Basic Hoods are economical and have a smaller overall height and depth than traditional hoods so they fit in areas with low ceilings or narrow aisles.
  • By-pass airflow design
  • Durable glacier white epoxy-coated 16 gauge steel exterior and interior resists chemical exposure
  • Vapor-proof incandescent lights
  • Vertical-rising tempered safety glass sash
  • Adjustable two-piece baffle
  • Available