Stampa…
LSD1 fluorometric drug discovery kit
Fornitore: ENZO LIFE SCIENCES
BMLAK5440001
ENZOBMLAK5440001EA 519 EUR
ENZOBMLAK5440001
LSD1 fluorometric drug discovery kit
Saggi Saggi cellulari
The LSD1 Fluorimetric Drug Discovery Kit provides human recombinant LSD1 and all reagents for measuring its activity in a sensitive, real-time fluorescent assay.
LSD1 catalyzed demethylation of the Histone H3 Dimethyl Lysine-4 Peptide (H3K4Me2 Peptide) produces hydrogen peroxide. A fluorescent signal is generated via the horseradish peroxidase (HRP) catalyzed reaction of the hydrogen peroxide with the CELLestial® Red Peroxidase Substrate. Although fluorescence detection will be more sensitive (Excitation in range of 530-570 nm; Emission ca. 590 nm), the CELLestial® Red peroxidation product may also be detected by absorbance at 563 nm.
Informazioni per l'ordine: Recombinant human LSD1 (50µg, >1000U)Histone H3 Dimethyl Lysine-4 Peptide SubstrateCELLestial® Red Peroxidase SubstrateHorseradish PeroxidaseH2O2 StandardTranylcypromine (LSD1 inhibitor)Black and clear ½-vol. 96-well platesDetailed instructions.