Stampa…
Annexin V-EGFP apoptosis detection kit
Fornitore: ENZO LIFE SCIENCES
ALX850253KI02 ALX850253KI02
ALEXALX850253KI02EA 0 EUR
ALEXALX850253KI02 ENZOALX850253KI02
Annexin V-EGFP apoptosis detection kit
Saggi Saggi cellulari
For the detection of apoptotic cells by flow cytometry.
EGFP (enhanced green fluorescent protein) has emerged as an ideal fluorochrome for FACS analysis and fluorescent microscopy as it offers a more intense signal than wild-type GFP. It has Ex(max): 488 nm and Em(max): 530 nm. The green signal produced makes EGFP ideal for dual staining with propidium iodide (PI).
Informazioni per l'ordine: Kit contains Annexin V-EGFP, propidium iodide solution, binding buffer and assay protocol.