Stampa…
ApoSENSOR™ Cell viability assay kit
Fornitore: ENZO LIFE SCIENCES
ALX850247KI01 ALX850247KI01
ALEXALX850247KI01EA 0 EUR
ALEXALX850247KI01 ENZOALX850247KI01
ApoSENSOR™ Cell viability assay kit
Saggi Saggi cellulari
The assay utilizes bioluminescent detection of the ATP level for a rapid screening of apoptosis and cell proliferation simultaneously in mammalian cells. The assay utilizes the enzyme luciferase to catalyze the formation of light from ATP and luciferin. The light can be measured using a luminometer or Beta Counter. The assay can be fully automatic for high throughput (10 seconds/sample) and is highly sensitive (detects 10-100 mammalian cells/well).
Informazioni per l'ordine: Kit includes Nucleotide Releasing Buffer, ATP Monitoring Enzyme, Enzyme Reconstitution Buffer and ATP.