Stampa…
Experimentation kits, PCR-based Alu-Human DNA typing
Fornitore: EDVOTEK
333
EDVO333EA 144 EUR
EDVO333
Experimentation kits, PCR-based Alu-Human DNA typing
Materiale didattico Materiali didattici di biologia Attività di apprendimento - PCR
Students use primers for a 300 base pair Alu insertion in chromosome 16 (PV92) to determine their own genotype. They can then compare their class results with others around the world over the internet.
  • For 25 students
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Informazioni per l'ordine: Kit includes instructions, proteinase K, PCR beads, control DNA and primers, microtubes, chelating agent, agarose,More Product Information