Stampa…
Experimentation kits, principles of enzyme catalysis
Fornitore: EDVOTEK
282
EDVO282EA 155 EUR
EDVO282
Experimentation kits, principles of enzyme catalysis
Materiale didattico Materiali didattici di biologia Biochemistry Learning Activities
This easy and safe experiment allows students to learn about enzyme catalysis, the nature of enzyme action, and protein structure-function relationship. Students will perform an enzyme assay and determine the rate of the enzymatic reaction.
  • For 10 lab groups
  • Requires 30 to 45 minutes
Also required: Visible wavelength spectrophotometer; 1 ml, 5 ml, and 10 ml pipettes; and linear graph paper.
Informazioni per l'ordine: Kit includes instructions, catalase solution, hydrogen peroxide, phosphate buffer, assay reagent, acidification solution, colour enhancer and developer.