Stampa…
Experimentation kits, isolation of E. coli Chromosomal DNA
Fornitore: EDVOTEK
203
EDVO203EA 167 EUR
EDVO203
Experimentation kits, isolation of E. coli Chromosomal DNA
Materiale didattico Materiali didattici di biologia Biochemistry Learning Activities
Isolation of high molecular weight chromosomal DNA is the first step in molecular cloning since it is the source of genes in cells. This experiment provides DNA extraction LyphoCells and reagents for isolating chromosomal DNA from E. coli. After spooling from solution, the DNA can be dissolved and analysed by agarose gel electrophoresis as an optional lab extension activity.
  • For 5 lab groups
  • Complete in 60 minutes
Also required: Water bath, 95 to 100% isopropanol. For optional electrophoresis: Electrophoresis tank, power supply, 5 to 50 µl adjustableMore Product Information