Stampa…
Prostaglandin Screening Library II
Fornitore: Cayman Chemical
10502-50
CAYM10502-50EA 2530 EUR
CAYM10502-50
Prostaglandin Screening Library II
Saggi Saggi cellulari
For D and E series prostaglandin analog screening.
  • Contains >60 compounds
  • Includes the ''D- and E-series'' prostaglandins
The Prostaglandin Screening Library II contains more than 60 ''D and E-series'' of prostaglandins in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.