Stampa…
Anti-Leptin Chicken Polyclonal Antibody
Fornitore: Biosensis
C-1510-500
BSENC-1510-500EA 321 EUR
BSENC-1510-500
Anti-Leptin Chicken Polyclonal Antibody
Anticorpi
Leptin is secreted by white adipocytes and functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. Leptin also has several endocrine functions and is involved in the regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing (Ref: Entrez Gene).
The antibody reacts with human leptin protein and may react with other speces leptin because of protein homology but is currently untested.
Application Information:
ELISA and WB. Suggested dilution of 1:1,000-1:2,000.More Product Information