Stampa…
Anti-Vimentin Chicken Polyclonal Antibody
Fornitore: Biosensis
C-1409-50
BSENC-1409-50EA 312 EUR
BSENC-1409-50
Anti-Vimentin Chicken Polyclonal Antibody
Anticorpi
Vimentins are class-III intermediate filaments specific to mesenchymal tissue. Vimentin is an important cytoskeletal component responsible for maintaining cell integrity and has a probable role in the intracellular transport of proteins such as lipoproteins between the nucleus and plasma membrane. Immunohistochemical staining for Vimentin is characteristic of sarcomas. 
The specificity of this antibody has been confirmed by WB. This antibody detects ~50 kDa Vimentin protein in multiple species. The antibody also reacts excellently in immunocytochemistry and immunhistochemistry.