Stampa…
NGF Rapid™ ELISA kit: human
Fornitore: Biosensis
BEK-2212-1P BEK-2212-2P
BSENBEK-2212-1PEA 398 EUR
BSENBEK-2212-1P BSENBEK-2212-2P
NGF Rapid™ ELISA kit: human
Saggi ELISA
The Biosensis NGF Rapid™ enzyme-linked immunosorbent assay (ELISA) kit is a sandwich ELISA that allows the quantification of human NGF in less than 3 hours in cell culture supernatants, serum, plasma (citrate) and brain extracts only if used as directed.
This NGF Rapid™ ELISA Kit is designed to measure human NGF and thus employs a recombinant human NGF standard approved by the World Health Organization (WHO). Due to a high degree of NGF sequence homology, the antibodies used in this kit will also detect NGF from other species including mouse and rat.
The purpose of this kit is the in vitro quantitative determination of NGF in samples such as serum, plasma, cell culture supernatants and brain extracts.
The antibodies used in this ELISA kit bind epitopes within the mature domain of the protein and therefore can recognize both the pro- and the mature form of NGF. However, internal validation data demonstrates <0,1% cross-reactivity (determined at 25 ng/ml, diluted in assay buffer) with human proNGF protein, suggesting the preferential quantification of mature NGF over full-length human proNGF. Note that accurate NGF quantification in human citrate-plasma requires the addition of Heterophilic Antibody Blocker provided in the kit, and available for purchase separately.
This kit consists of a pre-coated mouse monoclonal anti-NGF capture antibody, a biotinylated anti-NGF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a coloured reaction product which is directly proportional to the concentration of NGF present in samples and protein standards. A human NGF positive control (QC sample) is provided to assure consistent assay performance.
Please refer to the kit protocol for specific use instructions for each substrate application, in particular serum and plasma samples.
Informazioni per l'ordine: Each kit contains 1 or 2×96 pre-coated strip plates, protein standards, detection reagents, wash and sample buffers and detailed instructions.
Attenzione: For research use only. Not for diagnostic or clinical purposes.
Visualizza i codici
Specifiche
Environmentally Preferable
Target protein NGF
Primary antibody reactivity Human
Tipo di test Sandwich
Ospite
Formato Pre-coated
Sensibilità Typical limit of detection (LOD) for human NGF is 2 pg/ml determined as 150% of the blank value
Range di rilevamento 3,9 – 250 pg/ml
Reattività incrociata The antibodies used in this ELISA kit bind epitopes within the mature domain of the protein and therefore can recognise both the pro- and the mature form of NGF. However, human proNGF protein shows <0,1% cross-reactivity (determined at 25 ng/ml) allowing specific quantification of mature NGF. This NGF ELISA does not cross-react with brain derived neurotrophic factor (BDNF), neurotrophin-3 (NT-3), NT-4/5, glial cell line-derived neurotrophic factor (GDNF) and vascular endothelial growth factor (VEGF165) tested at 25 ng/ml. Due to a high degree of NGF sequence homology, the antibodies used in this kit will also detect NGF from other species including mouse and rat.