Stampa…
Anti-SIGLEC9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fornitore: Bioss
BS-9874R-A680
BOSSBS-9874R-A680EA 893 EUR
BOSSBS-9874R-A680
Anti-SIGLEC9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorpi
Two families of mammalian lectin-like adhesion molecules bind glycoconjugate ligands in a sialic acid-dependent manner: the selectins and the sialoadhesins. The sialic acid-binding immunoglobulin superfamily lectins, designated siglecs or sialoadhesins, are immunoglobulin superfamily members recognizing sialylated ligands. The common sialic acids of mammalian cells are N-acetyl-neuraminic acid (Neu5Ac) and N-glycolyl-neuraminic acid (Neu5Gc). Siglec-1 mediates local cell-cell interactions in lymphoid tissues and can be detected at contact points of macrophages with other macrophages, sinus-More Product Information