Stampa…

Anti-ARL6IP5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornitore: Bioss
BS-9706R-A555
BOSSBS-9706R-A555EA 893 EUR
BOSSBS-9706R-A555
Anti-ARL6IP5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpi
JWA is a four-transmembrane environmental responsive protein which binds to the CC chemokine recepor 5 (CCR5), a major co-receptor for human immunodeficiency virus (HIV). JWA is involved in environmental stress-responsive pathways in K562 cells, an erythroleukemia cell line derived from patients with chronic myeloid leukemia. Environmental stressors to K562 cells such as heat shock, a higher temperature than the ideal body temperature of the organism from which the cell line was derived, and oxidative stress, the production of oxygen-centered free radicals, regulate and increase the expres-More Product Information
Visualizza i codici

Specifiche

Simbolo dell'antigene ARL6IP5
Tipo di anticorpi ADP-Ribosylation Factor-Like 6 Interacting Protein 5
Environmentally Preferable
Sinomini antigene hp22|putative MAPK activating protein PM27|HSPC127|addicsin|ARL6IP5|jmx|JWA|ADP-ribosylation-like factor 6 interacting protein 5|PRAF3|glutamate transporter EEAC1-associated protein|PRA1 domain family 3|DERP11|GTRAP3-18|dermal papilla derived protein 11|cytoskeleton related vitamin A responsive protein
Tipo anticorpo Primary
Clonalità Policlonale
Coniugazione Alexa Fluor® 555
Reattività Human,Rat,Mouse
Ospite Rabbit
ID gene 10550
Isotipo IgG
ImmunoChimica Yes
Forma liquid
Purificazione Purified by Protein A
Concentrazione 1 μg/μl
Assorbimento incrociato No
Tampone di conservazione Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura di conservazione Store at –20 °C for 12 months
Temperatura di spedizione 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR