Stampa…
Anti-DHRS4 Rabbit Polyclonal Antibody
Fornitore: Bioss
BS-8264R
BOSSBS-8264REA 725 EUR
BOSSBS-8264R
Anti-DHRS4 Rabbit Polyclonal Antibody
Anticorpi
Reduces all-trans-retinal and 9-cis retinal. Can also catalyze the oxidation of all-trans-retinol with NADP as co-factor, but with much lower efficiency. Reduces alkyl phenyl ketones and alpha-dicarbonyl compounds with aromatic rings, such as pyrimidine-4-aldehyde, 3-benzoylpyridine, 4-benzoylpyridine, menadione and 4-hexanoylpyridine. Has no activity towards aliphatic aldehydes and ketones.Tissue specificity:Isoform 1 is predominantly expressed in normal cervix (at protein level). Isoform 4 is expressed in some neoplastic cervical tissues, but not in normal cervix (at protein level).More Product Information