Stampa…

Anti-ZMYND11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: Bioss
BS-7697R-HRP
BOSSBS-7697R-HRPEA 900 EUR
BOSSBS-7697R-HRP
Anti-ZMYND11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Chromatin reader that specifically recognizes and binds histone H3.3 trimethylated at 'Lys-36' (H3.3K36me3) and regulates RNA polymerase II elongation. Does not bind other histone H3 subtypes (H3.1 or H3.2) (By similarity). Colocalizes with highly expressed genes and functions as a transcription corepressor by modulating RNA polymerase II at the elongation stage. Acts as a tumor-suppressor by repressing a transcriptional program essential for tumor cell growth.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ZMYND11
Clonality: Polyclonal
Clone:
More Product Information
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR