Stampa…

Anti-DEDD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Fornitore: Bioss
BS-7062R-A680
BOSSBS-7062R-A680EA 893 EUR
BOSSBS-7062R-A680
Anti-DEDD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorpi
A scaffold protein that directs CASP3 to certain substrates and facilitates their ordered degradation during apoptosis. May also play a role in mediating CASP3 cleavage of KRT18. Regulates degradation of intermediate filaments during apoptosis. May play a role in the general transcription machinery in the nucleus and might be an important regulator of the activity of GTF3C3. Inhibits DNA transcription in vitro (By similarity).

Type: Primary
Antigen: DEDD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/318
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Others
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR