Stampa…

Anti-ELF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Fornitore: Bioss
BS-7042R-A488
BOSSBS-7042R-A488EA 893 EUR
BOSSBS-7042R-A488
Anti-ELF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpi
E74 ike Factor 1 is a transcription factor that may be required for the T-cell receptor mediated activation of HIV2 gene expression. It binds to two purine-rich motifs in the HIV-2 enhancer and transcriptionally activates the LYN and mouse BLK promoters.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ELF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR