Stampa…

Anti-AKT3 Rabbit Polyclonal Antibody

Fornitore: Bioss
BS-5209R
BOSSBS-5209REA 737 EUR
BOSSBS-5209R
Anti-AKT3 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR