Stampa…

Anti-PON1 Rabbit Polyclonal Antibody (Cy5.5®)

Fornitore: Bioss
BS-5107R-CY5.5
BOSSBS-5107R-CY5.5EA 900 EUR
BOSSBS-5107R-CY5.5
Anti-PON1 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorpi
Hydrolyzes the toxic metabolites of a variety of organophosphorus insecticides. Capable of hydrolyzing a broad spectrum of organophosphate substrates and lactones, and a number of aromatic carboxylic acid esters. Mediates an enzymatic protection of low density lipoproteins against oxidative modification and the consequent series of events leading to atheroma formation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PON1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Specifiche

Simbolo dell'antigene PON1
Environmentally Preferable
Tipo di anticorpi Paraoxonase 1
Sinomini antigene esterase A|PON|paraoxonase 1|ESA|PON1
Tipo anticorpo Primary
Clonalità Policlonale
Coniugazione Cy5.5®
Reattività Human,Rat,Mouse
Ospite Rabbit
ID gene 5444
Isotipo IgG
ImmunoChimica Yes
Forma liquid
Purificazione Purified by Protein A
Concentrazione 1 μg/μl
Assorbimento incrociato No
Tampone di conservazione Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura di conservazione Store at 4°C for 12 months
Temperatura di spedizione 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR