Stampa…

Anti-PON1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Fornitore: Bioss
BS-5107R-A680
BOSSBS-5107R-A680EA 890 EUR
BOSSBS-5107R-A680
Anti-PON1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorpi
hydrolyses the toxic metabolites of a variety of organophosphorus insecticides. Capable of hydrolyzing a broad spectrum of organophosphate substrates and lactones, and a number of aromatic carboxylic acid esters. Mediates an enzymatic protection of low density lipoproteins against oxidative modification and the consequent series of events leading to atheroma formation.

Type: Primary
Antigen: PON1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/355
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR