Stampa…

Anti-ND5 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BS-3952R-CY7
BOSSBS-3952R-CY7EA 909 EUR
BOSSBS-3952R-CY7
Anti-ND5 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ND5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR