Stampa…

Anti-SLCO2B1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Fornitore: Bioss
BS-3913R-A488
BOSSBS-3913R-A488EA 901 EUR
BOSSBS-3913R-A488
Anti-SLCO2B1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpi
Mediates the Na(+)-independent transport of organic anions such as taurocholate, the prostaglandins PGD2, PGE1, PGE2, leukotriene C4, thromboxane B2 and iloprost.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLCO2B1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR