Stampa…
Anti-INPPL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fornitore: Bioss
BS-3404R-A488
BOSSBS-3404R-A488EA 921 EUR
BOSSBS-3404R-A488
Anti-INPPL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpi
The steady state of protein tyrosyl phosphorylation in cells is regulated by the opposing action of tyrosine kinases and protein tyrosine phosphatases (PTPs). Several groups have independently identified a non transmembrane PTP, designated SHPTP1 (also known as PTP1C, HCP and SHP), which is primarily expressed in hematopoietic cells and characterized by the presence of two SH2 domains N terminal to the PTP domain. A second and much more widely expressed PTP with SH2 domains, SHPTP2 (also designated PTP1D and Syp), has been identified. SHP2 is a protein tyrosine phosphatase that is widelyMore Product Information