Stampa…

Anti-BMX Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornitore: Bioss
BS-3132R-FITC
BOSSBS-3132R-FITCEA 929 EUR
BOSSBS-3132R-FITC
Anti-BMX Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
This gene encodes a non-receptor tyrosine kinase belonging to the Tec kinase family. The protein contains a PH-like domain, which mediates membrane targeting by binding to phosphatidylinositol 3,4,5-triphosphate (PIP3), and a SH2 domain that binds to tyrosine-phosphorylated proteins and functions in signal transduction. The protein is implicated in several signal transduction pathways including the Stat pathway, and regulates differentiation and tumorigenicity of several types of cancer cells. Multiple alternatively spliced variants, encoding the same protein, have been identified.[providedMore Product Information
Visualizza i codici

Specifiche

Simbolo dell'antigene BMX
Environmentally Preferable
Tipo di anticorpi BMX non-receptor tyrosine kinase
Sinomini antigene Bone marrow kinase BMX|NTK38|ETK|Epithelial and endothelial tyrosine kinase|Cytoplasmic tyrosine-protein kinase BMX
Tipo anticorpo Primary
Clonalità Policlonale
Coniugazione FITC (Fluoresceina Isotiocionato)
Reattività Human,Rat,Mouse
Ospite Rabbit
ID gene 660
Isotipo IgG
ImmunoChimica Yes
Modifica phospho-Tyr40
Forma liquid
Purificazione Purified by Protein A
Concentrazione 1 μg/μl
Assorbimento incrociato No
Tampone di conservazione Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura di conservazione Store at 4°C for 12 months
Temperatura di spedizione 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR