Stampa…

Anti-CLDN4 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BS-2790R-CY7
BOSSBS-2790R-CY7EA 909 EUR
BOSSBS-2790R-CY7
Anti-CLDN4 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact made up of network of strands that act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLDN4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR