Stampa…
Anti-PAX5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fornitore: Bioss
BS-1861R-A350
BOSSBS-1861R-A350EA 893 EUR
BOSSBS-1861R-A350
Anti-PAX5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorpi
The PAX5 gene is a member of the paired box (PAX) family of transcription factors. The central feature of this gene family is a novel, highly conserved DNA-binding motif, known as the paired box. The PAX proteins are important regulators in early development, and alterations in the expression of their genes are thought to contribute to neoplastic transformation. The PAX5 gene encodes the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5More Product Information