Stampa…
Anti-C1orf159 Rabbit Polyclonal Antibody (Cy3®)
Fornitore: Bioss
BS-15031R-CY3
BOSSBS-15031R-CY3EA 900 EUR
BOSSBS-15031R-CY3
Anti-C1orf159 Rabbit Polyclonal Antibody (Cy3®)
Anticorpi
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. TheMore Product Information