Stampa…
Anti-FLT3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornitore: Bioss
BS-13184R-HRP
BOSSBS-13184R-HRPEA 929 EUR
BOSSBS-13184R-HRP
Anti-FLT3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Tyrosine-protein kinase that acts as cell-surface receptor for the cytokine FLT3LG and regulates differentiation, proliferation and survival of hematopoietic progenitor cells and of dendritic cells. Promotes phosphorylation of SHC1 and AKT1, and activation of the downstream effector MTOR. Promotes activation of RAS signaling and phosphorylation of downstream kinases, including MAPK1/ERK2 and/or MAPK3/ERK1. Promotes phosphorylation of FES, FER, PTPN6/SHP, PTPN11/SHP-2, PLCG1, and STAT5A and/or STAT5B. Activation of wild-type FLT3 causes only marginal activation of STAT5A or STAT5B. MutationsMore Product Information