Stampa…

Anti-NOS3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornitore: Bioss
BS-13074R-A555
BOSSBS-13074R-A555EA 898 EUR
BOSSBS-13074R-A555
Anti-NOS3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpi
eNOS is a calcium/calmodulin dependent enzyme which undergoes several post translational modifications, including acylation with myristate and palmitate, and phosphorylation on numerous residues. As with the other members of the NOS family, eNOS derives the diffusible multifunctional second messenger NO from L arginine through a series of reactions in which L citrulline is an intermediate. eNOS plays an important role in controlling vascular tone, platelet aggregation, and cardiac myocyte function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOS3
Clonality:More Product Information
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR