Stampa…

Anti-APOA1BP Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BS-12491R-CY7
BOSSBS-12491R-CY7EA 900 EUR
BOSSBS-12491R-CY7
Anti-APOA1BP Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Apolipoproteins are protein components of plasma lipoproteins. Apolipoprotein A-I (ApoA1) promotes cholesterol efflux from tissues to the liver for excretion. ApoA1 is the major protein component of high density lipoprotein (HDL) in the plasma. It can function as a cofactor for lecithin cholesterolacyltransferase, which is responsible for the formation of most plasma cholesteryl esters. AI-BP (Apolipoprotein A-I-binding protein), also known as YjeF N-terminal domain-containing protein 1, is a 288 amino acid secreted protein that binds ApoA1, ApoA2 and HDL. Individuals with impaired renalMore Product Information
Visualizza i codici

Specifiche

Simbolo dell'antigene APOA1BP
Tipo di anticorpi Apolipoprotein A-I Binding Protein
Environmentally Preferable
Sinomini antigene YJEFN1|AIBP
Tipo anticorpo Primary
Clonalità Policlonale
Coniugazione Cy7®
Reattività Human,Rat,Mouse
Ospite Rabbit
ID gene 128240
Isotipo IgG
ImmunoChimica Yes
Forma liquid
Purificazione Purified by Protein A
Concentrazione 1 μg/μl
Assorbimento incrociato No
Tampone di conservazione Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura di conservazione Store at 4°C for 12 months
Temperatura di spedizione 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR