Stampa…
Anti-GABARE Rabbit Polyclonal Antibody (Cy7®)
Fornitore: Bioss
BS-12081R-CY7
BOSSBS-12081R-CY7EA 900 EUR
BOSSBS-12081R-CY7
Anti-GABARE Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct. Members of the GABAA receptor family include GABAA R Alpha 1-6, GABAA R Beta 1-3,More Product Information