Stampa…
Anti-LALBA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornitore: Bioss
BS-11131R-FITC
BOSSBS-11131R-FITCEA 900 EUR
BOSSBS-11131R-FITC
Anti-LALBA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Alpha-lactalbumin is the B protein of lactose synthetase secreted by the mammary epithelial cells. It is a potent Ca2+-elevating and apoptosis-inducing agent with broad, yet selective, cytotoxic activity. Multimeric ?lactalbumin has been shown to kill all transformed, embryonic and lymphoid cells tested, but not mature epithelial elements. This suggests that milk contributes to mucosal immunity not only by furnishing antimicrobial molecules but also by policing the function of lymphocytes and epithelium. ?lactalbumin may be helpful in discovering the site of origin of metastatic breastMore Product Information