Stampa…
Anti-CD3E Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fornitore: Bioss
BS-0765R-A488
BOSSBS-0765R-A488EA 901 EUR
BOSSBS-0765R-A488
Anti-CD3E Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpi
The protein encoded by this gene is the CD3-epsilon polypeptide, which together with CD3-gamma, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. The epsilon polypeptide plays an essential role in T-cell development. Defects in this gene cause immunodeficiency. This gene has also beenMore Product Information