Stampa…

Anti-c-Kit Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Fornitore: Bioss
BS-0672R-A680
BOSSBS-0672R-A680EA 899 EUR
BOSSBS-0672R-A680
Anti-c-Kit Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorpi
Tyrosine-protein kinase that acts as cell-surface receptor for the cytokine KITLG/SCF and plays an essential role in the regulation of cell survival and proliferation, hematopoiesis, stem cell maintenance, gametogenesis, mast cell development, migration and function, and in melanogenesis. In response to KITLG/SCF binding, KIT can activate several signaling pathways. Phosphorylates PIK3R1, PLCG1, SH2B2/APS and CBL. Activates the AKT1 signaling pathway by phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase. Activated KIT also transmits signals via GRB2 andMore Product Information
Visualizza i codici

Specifiche

Environmentally Preferable
Tipo di anticorpi c-Kit
Sinomini antigene SCFR|v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog|p145 c-kit|CD117|Piebald trait protein|PBT|KIT|Tyrosine-protein kinase Kit|C-Kit|Proto-oncogene c-Kit|Mast/stem cell growth factor receptor Kit
Tipo anticorpo Primary
Clonalità Policlonale
Coniugazione Alexa Fluor® 680
Reattività Human,Rat,Mouse
Ospite Rabbit
ID gene 3815
Isotipo IgG
Western blot Yes
ImmunoChimica Yes
ImmunoFluorescenza Yes
Citometria a flusso Yes
Modifica Unmodified
Epitopo 901-976/976
Immunogeno KLH conjugated synthetic peptide derived from human CD117/c-kit
Forma Liquid
Peso molecolare 105 kDa
Purificazione Purified by Protein A
Concentrazione 1 µg/µl
Tampone di conservazione Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Temperatura di conservazione Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Temperatura di spedizione 4 °C
Applicazioni testate ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR