Stampa…

Acido 5,5'-ditiobis (2-nitrobenzoico) (Reattivo di Ellman), Molecular Biology Grade

Fornitore: G Biosciences
Attenzione

BC87
BC87EA 288 EUR
BC87
Acido 5,5'-ditiobis (2-nitrobenzoico) (Reattivo di Ellman), Molecular Biology Grade
5,5'-Ditiobis(acido 2-nitrobenzoico) (Reattivo di Ellman)

Ellman's reagent is a versatile, water-soluble compound for quantifying free sulfhydryl groups in solution. It reacts with a free sulfhydryl group to yield a mixed disulfide and 2-nitro-5-thiobenzoic acid (NTB), a measurable yellow colored product at 412nm.
Ellman's reagent is very useful as a free sulfhydryl assay reagent due to its high specificity for -SH groups at neutral pH, high molar extinction coefficient and short reaction time.


Yellow crystalline powder that is used for the colorimetric determination of thiol groups in biological samples. DTNB is fairly soluble in water.
Store at 2-8°C and protect from light and humidity.


Applications: Quantification of free sulfhydryl residues

Formula: C₁₄H₈N₂O₈S₂
Punto di fusione: 240…244 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00007140
CAS Number: 69-78-3
EINECS: 200-714-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR