Stampa…

Acqua, Molecular Grade DNase-, RNase-free

Fornitore: G Biosciences

786-72C 786-73C 786-292 786-293
786-72CEA 210 EUR
786-72C 786-73C 786-292 786-293
Acqua, Molecular Grade DNase-, RNase-free
Acqua

Molecular Grade Water is ultrapure water that certified RNase, DNase and Protease free, is suitable for use in molecular biology applications, which demand a high quality of water and assurance that the water is free from nucleases and protease contamination. No toxic agents, such as DEPC, are used in the manufacturing of Molecular Grade Water, eliminating DEPC interference of enzymatic reactions.


  • Each lot is quality tested for the absence of RNase, DNase and protease contamination


For use in molecular biology applications, PCR reactions, 2D electrophoresis and mass spectrometry.

Formula: H₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR